Assay Details
Gene Symbol: PEG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005596576.3 | XP_005596633.2 | 826 | 81 | ||
XM_005596577.3 | XP_005596634.2 | 749 | 81 | |||
XM_014730904.2 | XP_014586390.2 | 1142 | 81 | |||
XM_023650574.1 | XP_023506342.1 | 1231 | 81 | |||
XM_023650575.1 | XP_023506343.1 | 2186 | 81 | |||
XM_023650576.1 | XP_023506344.1 | 1362 | 81 | |||
XM_023650577.1 | XP_023506345.1 | 1435 | 81 | |||
XM_023650578.1 | XP_023506346.1 | 916 | 81 | |||
XM_023650579.1 | XP_023506347.1 | 1871 | 81 | |||
XM_023650580.1 | XP_023506348.1 | 839 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
