Assay Details
Gene Symbol: SH3BGRL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023650692.1 | XP_023506460.1 | 370 | 96 | ||
XM_023650693.1 | XP_023506461.1 | 522 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
