Assay Details
Gene Symbol: DDO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001916542.4 | XP_001916577.2 | 1133 | 69 | ||
XM_014734560.2 | XP_014590046.1 | 548 | 69 | |||
XM_014734561.2 | XP_014590047.1 | 499 | 69 | |||
XM_014734564.2 | XP_014590050.1 | 548 | 69 | |||
XM_014734567.2 | XP_014590053.1 | 504 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
