Assay Details
Gene Symbol: RHOC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001917242.5 | XP_001917277.2 | 532 | 81 | ||
XM_023641407.1 | XP_023497175.1 | 554 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
