Assay Details
Gene Symbol: TAF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493837.5 | XP_001493887.1 | 299 | 85 | ||
XM_023641491.1 | XP_023497259.1 | 340 | 85 | |||
XM_023641492.1 | XP_023497260.1 | 448 | 85 | |||
GenBank mRNA | JL615997.1 | - | 3 - 4 | 230 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
