Assay Details
Gene Symbol: EXTL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005610394.3 | XP_005610451.1 | 1209 | 88 | ||
XM_023641541.1 | XP_023497309.1 | 937 | 88 | |||
XM_023641542.1 | XP_023497310.1 | 930 | 88 | |||
XM_023641543.1 | XP_023497311.1 | 811 | 88 | |||
XM_023641544.1 | XP_023497312.1 | 733 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
