Assay Details
Gene Symbol: ZUP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504142.6 | XP_001504192.1 | 1722 | 77 | ||
XM_023650967.1 | XP_023506735.1 | 1723 | 77 | |||
XM_023650968.1 | XP_023506736.1 | 1432 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
