Assay Details
Gene Symbol: CYBC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023651706.1 | XP_023507474.1 | 286 | 112 | ||
XM_023651707.1 | XP_023507475.1 | 1023 | 112 | |||
XM_023651709.1 | XP_023507477.1 | 286 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
