Assay Details
Gene Symbol: OGFOD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023651704.1 | XP_023507472.1 | 440 | 66 | ||
XM_023651705.1 | XP_023507473.1 | 440 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
