Assay Details
Gene Symbol: RAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023651746.1 | XP_023507514.1 | 364 | 59 | ||
XM_023651747.1 | XP_023507515.1 | 355 | 59 | |||
XM_023651748.1 | XP_023507516.1 | 319 | 59 | |||
XM_023651749.1 | XP_023507517.1 | 360 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
