Assay Details
Gene Symbol: PYCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001487957.6 | XP_001488007.1 | 259 | 59 | ||
XM_014737687.2 | XP_014593173.1 | 260 | 59 | |||
XM_023651759.1 | XP_023507527.1 | 217 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
