Assay Details
Gene Symbol: MCRIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488427.5 | XP_001488477.1 | 75 | 69 | ||
XM_005597054.3 | XP_005597111.1 | 125 | 69 | |||
XM_005597056.3 | XP_005597113.1 | 425 | 69 | |||
XM_014737752.2 | XP_014593238.1 | 453 | 69 | |||
XM_023651772.1 | XP_023507540.1 | 406 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001488427.5, XM_005597056.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
