Assay Details
Gene Symbol: C1QTNF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005597090.3 | XP_005597147.1 | 407 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005597090.3 transcripts.
Set Membership: |
![]() ![]() |
