Assay Details
Gene Symbol: C1QTNF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005597089.3 | XP_005597146.1 | 561 | 97 | ||
XM_005597090.3 | XP_005597147.1 | 565 | 97 | |||
XM_023651897.1 | XP_023507665.1 | 247 | 97 |
