Assay Details
Gene Symbol: METTL23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491460.5 | XP_001491510.2 | 391 | 122 | ||
XM_005597115.3 | XP_005597172.1 | 1393 | 122 | |||
XM_005597116.3 | XP_005597173.2 | 582 | 122 | |||
XM_005597117.3 | XP_005597174.2 | 691 | 122 | |||
XM_023651954.1 | XP_023507722.1 | 902 | 122 | |||
GenBank mRNA | JL616453.2 | - | 2 - 3 | 307 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() |
