Assay Details
Gene Symbol: METTL23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491460.5 | XP_001491510.2 | 485 | 75 | ||
XM_005597115.3 | XP_005597172.1 | 1487 | 75 | |||
XM_005597116.3 | XP_005597173.2 | 676 | 75 | |||
XM_005597117.3 | XP_005597174.2 | 785 | 75 | |||
XM_023651954.1 | XP_023507722.1 | 996 | 75 | |||
GenBank mRNA | JL616453.2 | - | 3 - 4 | 401 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
