Assay Details
Gene Symbol: NT5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023652055.1 | XP_023507823.1 | 425 | 72 | ||
XM_023652056.1 | XP_023507824.1 | 405 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
