Assay Details
Gene Symbol: ATP5PD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001496541.4 | XP_001496591.1 | 517 | 142 | ||
XM_005597174.3 | XP_005597231.1 | 499 | 142 | |||
XM_005597175.3 | XP_005597232.1 | 495 | 142 | |||
XM_005597176.3 | XP_005597233.1 | 451 | 142 | |||
XM_023652058.1 | XP_023507826.1 | 436 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() |
