Assay Details
Gene Symbol: RPL38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003362519.4 | XP_003362567.2 | 237 | 93 | ||
XM_005597217.3 | XP_005597274.1 | 76 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005597217.3 transcripts.
Set Membership: |
![]() ![]() |
