Assay Details
Gene Symbol: POLG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495189.5 | XP_001495239.3 | 1964 | 64 | ||
XM_005597266.3 | XP_005597323.2 | 1964 | 64 | |||
XM_023652205.1 | XP_023507973.1 | 1964 | 64 | |||
XM_023652206.1 | XP_023507974.1 | 1964 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
