Assay Details
Gene Symbol: MILR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005597269.2 | XP_005597326.1 | 711 | 93 | ||
XM_014739278.2 | XP_014594764.1 | 711 | 93 | |||
XM_014739290.2 | XP_014594776.1 | 711 | 93 | |||
XM_014739297.2 | XP_014594783.1 | 591 | 93 | |||
XM_014739307.2 | XP_014594793.1 | 441 | 93 | |||
XM_023652208.1 | XP_023507976.1 | 711 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
