Assay Details
Gene Symbol: CD79B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005597902.3 | XP_005597959.2 | 624 | 78 | ||
XM_023652220.1 | XP_023507988.1 | 627 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
