Assay Details
Gene Symbol: CD300LG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495705.4 | XP_001495755.2 | 542 | 77 | ||
XM_005597351.2 | XP_005597408.1 | 848 | 77 | |||
XM_005597352.2 | XP_005597409.1 | 746 | 77 | |||
XM_014739871.2 | XP_014595357.1 | 824 | 77 | |||
XM_014739879.2 | XP_014595365.1 | 848 | 77 | |||
XM_014739883.2 | XP_014595369.1 | 848 | 77 | |||
XM_014739885.2 | XP_014595371.1 | 644 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
