Assay Details
Gene Symbol: CNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005597392.3 | XP_005597449.1 | 698 | 82 | ||
XM_014740234.2 | XP_014595720.1 | 826 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
