Assay Details
Gene Symbol: WIPF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014740606.2 | XP_014596092.1 | 305 | 72 | ||
XM_023652565.1 | XP_023508333.1 | 308 | 72 | |||
XM_023652566.1 | XP_023508334.1 | 259 | 72 | |||
XM_023652567.1 | XP_023508335.1 | 389 | 72 | |||
XM_023652568.1 | XP_023508336.1 | 155 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
