Assay Details
Gene Symbol: PPP1R1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498077.6 | XP_001498127.3 | 566 | 67 | ||
XM_023652600.1 | XP_023508368.1 | 725 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
