Assay Details
Gene Symbol: PNPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501917.4 | XP_001501967.2 | 756 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
