Assay Details
Gene Symbol: TTLL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023652681.1 | XP_023508449.1 | 4216 | 79 | ||
XM_023652682.1 | XP_023508450.1 | 3341 | 79 | |||
XM_023652683.1 | XP_023508451.1 | 2228 | 79 | |||
XM_023652684.1 | XP_023508452.1 | 2182 | 79 | |||
XM_023652685.1 | XP_023508453.1 | 1729 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
