Assay Details
Gene Symbol: ABR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504257.6 | XP_001504307.4 | 3224 | 71 | ||
XM_001504260.6 | XP_001504310.3 | 1986 | 71 | |||
XM_005597643.3 | XP_005597700.1 | 468 | 71 | |||
XM_023653176.1 | XP_023508944.1 | 3224 | 71 | |||
XM_023653177.1 | XP_023508945.1 | 2097 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
