Assay Details
Gene Symbol: YWHAE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504287.5 | XP_001504337.1 | 181 | 80 | ||
XM_005597648.3 | XP_005597705.1 | 140 | 80 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: YWHAE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504287.5 | XP_001504337.1 | 181 | 80 | ||
XM_005597648.3 | XP_005597705.1 | 140 | 80 |