Assay Details
Gene Symbol: METTL16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005597657.3 | XP_005597714.1 | 745 | 64 | ||
XM_014727919.2 | XP_014583405.1 | 798 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
