Assay Details
Gene Symbol: SPATA22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504720.3 | XP_001504770.1 | 390 | 73 | ||
XM_023653268.1 | XP_023509036.1 | 664 | 73 | |||
XM_023653269.1 | XP_023509037.1 | 393 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
