Assay Details
Gene Symbol: XAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005597683.3 | XP_005597740.1 | 232 | 69 | ||
XM_014728082.2 | XP_014583568.1 | 230 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
