Assay Details
Gene Symbol: FGF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005597747.3 | XP_005597804.1 | 1741 | 80 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FGF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005597747.3 | XP_005597804.1 | 1741 | 80 |