Assay Details
Gene Symbol: AURKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504814.4 | XP_001504864.1 | 1008 | 71 | ||
XM_014728662.1 | XP_014584148.1 | 401 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
