Assay Details
Gene Symbol: SCO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005597790.3 | XP_005597847.2 | 820 | 72 | ||
GenBank mRNA | JL619483.1 | - | 3 - 4 | 419 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
