Assay Details
Gene Symbol: MAP2K4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023653527.1 | XP_023509295.1 | 952 | 53 | ||
XM_023653528.1 | XP_023509296.1 | 918 | 53 | |||
XM_023653529.1 | XP_023509297.1 | 3211 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() |
