Assay Details
Gene Symbol: ATPAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489182.4 | XP_001489232.1 | 495 | 68 | ||
XM_005597826.3 | XP_005597883.1 | 613 | 68 | |||
XM_005597827.3 | XP_005597884.1 | 613 | 68 | |||
XM_005597828.3 | XP_005597885.1 | 496 | 68 | |||
XM_005597829.3 | XP_005597886.1 | 613 | 68 | |||
XM_005597830.3 | XP_005597887.1 | 451 | 68 | |||
XM_005597831.3 | XP_005597888.1 | 613 | 68 | |||
XM_023653663.1 | XP_023509431.1 | 613 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
