Assay Details
Gene Symbol: TRAF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023653892.1 | XP_023509660.1 | 1063 | 64 | ||
XM_023653893.1 | XP_023509661.1 | 1212 | 64 | |||
XM_023653894.1 | XP_023509662.1 | 987 | 64 | |||
XM_023653895.1 | XP_023509663.1 | 918 | 64 | |||
XM_023653896.1 | XP_023509664.1 | 2795 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
