Assay Details
Gene Symbol: SPI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005598097.3 | XP_005598154.1 | 244 | 90 | ||
GenBank mRNA | JN979559.1 | - | 1 - 2 | 143 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
