Assay Details
Gene Symbol: SPI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491380.5 | XP_001491430.1 | 336 | 93 | ||
XM_005598097.3 | XP_005598154.1 | 339 | 93 | |||
GenBank mRNA | JN979559.1 | - | 2 - 3 | 238 | 93 | |
JN979560.1 | - | 2 - 3 | 235 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
