Assay Details
Gene Symbol: SPI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491380.5 | XP_001491430.1 | 523 | 101 | ||
XM_005598097.3 | XP_005598154.1 | 526 | 101 | |||
GenBank mRNA | JN979559.1 | - | 3 - 4 | 425 | 101 | |
JN979560.1 | - | 3 - 4 | 422 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
