Assay Details
Gene Symbol: CLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497108.5 | XP_001497158.2 | 749 | 71 | ||
XM_005598149.3 | XP_005598206.1 | 779 | 71 | |||
XM_005598150.3 | XP_005598207.1 | 883 | 71 | |||
XM_023654087.1 | XP_023509855.1 | 762 | 71 | |||
XM_023654088.1 | XP_023509856.1 | 777 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
