Assay Details
Gene Symbol: TCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005598191.3 | XP_005598248.1 | 795 | 112 | ||
GenBank mRNA | JL625492.1 | - | 2 - 3 | 308 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() |
