Assay Details
Gene Symbol: CYB561A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005598223.3 | XP_005598280.1 | 1015 | 67 | ||
XM_005598224.3 | XP_005598281.1 | 1106 | 67 | |||
XM_005598225.2 | XP_005598282.1 | 820 | 67 | |||
XM_005598226.3 | XP_005598283.1 | 1015 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
