Assay Details
Gene Symbol: SLC22A24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502945.3 | XP_001502995.1 | 1185 | 137 | ||
XM_014729635.2 | XP_014585121.1 | 1185 | 137 |

More Information
Set Membership: |
![]() ![]() |
