Assay Details
Gene Symbol: ZNF644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493570.6 | XP_001493620.1 | 4130 | 79 | ||
XM_023641680.1 | XP_023497448.1 | 4008 | 79 | |||
XM_023641681.1 | XP_023497449.1 | 3402 | 79 | |||
XM_023641682.1 | XP_023497450.1 | 4006 | 79 | |||
XM_023641683.1 | XP_023497451.1 | 3400 | 79 | |||
XM_023641684.1 | XP_023497452.1 | 3457 | 79 |
