Assay Details
Gene Symbol: ZNF644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493570.6 | XP_001493620.1 | 4735 | 93 | ||
GenBank mRNA | JL618455.1 | - | 1 - 2 | 608 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
