Assay Details
Gene Symbol: HS2ST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494558.5 | XP_001494608.2 | 1000 | 73 | ||
XM_005610449.3 | XP_005610506.1 | 1005 | 73 | |||
GenBank mRNA | JL632943.1 | - | 1 - 2 | 98 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
