Assay Details
Gene Symbol: GIPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014740030.2 | XP_014595516.2 | 1490 | 95 | ||
XM_023641792.1 | XP_023497560.1 | 1498 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
